Honeywell Safety Products™ Sperianl™ One-Fit™ N95 Particulate Respirators
Universal sized disposable respirators fit most face shapes and sizes
Manufacturer:  Honeywell Safety Products™ 14110444
 View more versions of this product
Catalog No. 29-635-9660
Description
Description
  • Electrostatic nonwoven polypropylene and polyester materials; lightweight construction allows for longer wear time and less fatigue
  • Durable filtration media ensures filtration efficiency of 95% against nonoil airborne particles
  • Protection against airborne pathogens and M. tuberculosis as well as certain airborne particulates and pandemic diseases such as H5N1 Avian Flu and SARs
W1400 Flat Fold Mask
  • Bonded wire nose clip with open cell foam nose seal
  • Low-profile design offers compatibility with spectacles
NBW95 and NBW95V Form Fitted Masks
  • Strong and rigid outer shell prevents mask from collapse in high humidity or moist environments
  • Molded nose bridge fits comfortably and is compatible with spectacles
Specifications
Specifications
NIOSH
Particulate Respirator
White
Yes
Protects against airborne particulate and pandemic disease
Electrostatic Nonwoven Polypropylene and Polyester
Each
Lightweight
White
N95 Filter
Yes
Universal
Documents