Please login to your online account to display your discounted pricing

North™ NorthFlex Red™ Foamed PVC Palm Coated Gloves

Light and comfortable general purpose gloves that offer high abrasion resistance and exceptional grip

$3.31 - $3.31

Specifications

Cleaning Procedure Launderable
Coated Area Full palm, finger
Color Red
Material PVC (Polyvinyl Chloride)
For Use With (Application) agriculture, construction, manufacturing, municipal services, Steel and Metals, utilities
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-166-666

 
honeywell safety products
NF11/8M
Medium Pair for $3.31
Case of 144 Pair for $424.80

19-166-667

 
honeywell safety products
NF11/9L
Large Pair for $3.31
Case of 144 Pair for $424.80

19-166-665

 
honeywell safety products
NF11/7S
Small Pair for $3.31
Case of 144 Pair for $424.80

19-166-668

 
honeywell safety products
NF11/10XL
X-Large Pair for $3.31
Case of 144 Pair for $424.80
Description & Specifications

Specifications

Cleaning Procedure Launderable
Coated Area Full palm, finger
Color Red
Material PVC (Polyvinyl Chloride)
For Use With (Application) agriculture, construction, manufacturing, municipal services, Steel and Metals, utilities
Glove Cuff Style Knit

Ideal for applications where workers handle both wet and dry objects and need moderate protection from mechanical hazards.

  • Advanced 15 gauge seamless nylon liners are stretchy and form fitting, with a snug knit-wrist, and provide minimal resistance to hand movement for exceptional wearer comfort
  • Foamed PVC coatings, are soft and flexible enough to provide a degree of shock and vibration absorption, but are tough enough to offer abrasion resistance comparable to polyurethane
  • In addition, foamed PVC has an open surface texture that clears liquids from surfaces and enables a very firm wet-grip
  • The Red-X™ features an extended knuckle coating, for additional wearer protection
  • The back of the glove is un-coated, for ventilation and wearer comfort
  • Launderable for added value and longer life
  • Cuff over-stitching is color-coded by size to aid sorting after laundering