Honeywell™ North™ NorthFlex Light Task and Light Task Plus II™ Gloves
PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life
Manufacturer:  Honeywell™ NF158M
 View more versions of this product
Catalog No. 29-635-7293
Description
Description
North NorthFlex Light Task Polyurethane Coated Nylon Gloves
  • Not cut-resistant
  • Polyurethane palm coating provides excellent grip and longer life
  • Uncoated back design allows gloves to breathe for more comfort
  • 15 gauge nylon liner has minimal shrinkage when laundered
  • Cuff over-stitching is color-coded by size to aid sorting after laundering
Specifications
Specifications
Safety
Level 1
Sandy
automotive assembly, electronics, general maintenance work, light assembly, manufacturing, small part handling
Nylon
White
Palm
Launderable
Knit
Polyurethane
Medium
Green
Documents