Honeywell™ North™ NorthFlex Light Task and Light Task Plus II™ Gloves
PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life
Manufacturer:  Honeywell™ NF157S*
 View more versions of this product
Catalog No. 29-635-7292
Description
Description
North NorthFlex Light Task Polyurethane Coated Nylon Gloves
  • Not cut-resistant
  • Polyurethane palm coating provides excellent grip and longer life
  • Uncoated back design allows gloves to breathe for more comfort
  • 15 gauge nylon liner has minimal shrinkage when laundered
  • Cuff over-stitching is color-coded by size to aid sorting after laundering
Specifications
Specifications
Palm
Sandy
automotive assembly, electronics, general maintenance work, light assembly, manufacturing, small part handling
Nylon
White
Launderable
Knit
Polyurethane
Small
Red