Please login to your online account to display your discounted pricing

Honeywell North™ NorthFlex Light Task Plus II™ Gloves

PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life

$13.30 - $142.43

Specifications

Color White
Cut Resistance Rating Level 2
Glove Cuff Style Knit
View More Specs
Products
Catalog Number Mfr. No. Coated Area Liner Material Material Size Price Quantity    

19-150-1002

 
honeywell
NFD15/7S
Palm, wrist Dyneema™ Fiber Nitrile, Polyurethane Small Dozen of 12 Each for $142.43

29-635-7328

 
honeywell
NFD158M
Palm, wrist Dyneema™ Fiber Nitrile, Polyurethane Medium Pair for $13.30
Dozen of 12 Pair for $154.80

19-150-1004

 
honeywell
NFD15/9L
Palm, wrist Dyneema™ Fiber Nitrile, Polyurethane Large Dozen of 12 Each for $142.43

19-150-1005

 
honeywell
NFD15/10XL
Palm, wrist Dyneema™ Fiber Nitrile, Polyurethane X-Large Dozen of 12 Each for $142.43
Description & Specifications

Specifications

Color White
Cut Resistance Rating Level 2
Glove Cuff Style Knit

North NorthFlex Light Task II Polyurethane Coated Dyneema Gloves

  • PU (polyurethane) palm coating is light for comfort and maximum tactility, but provides high abrasion and puncture resistance
  • Glove conforms to the shape of the hand and provides minimal resistance to hand movement, thus greatly reducing fatigue
  • The back of the glove is un-coated, for ventilation and wearer comfort
  • Contains no reaction causing, natural rubber proteins
  • Cuff over-stitching is color-coded by size to aid sorting after laundering

Complies with USDA and FDA regulations, 21 CFR, for use in food processing. ANSI cut level 1 and EN388 cut level 2 performance.