Honeywell™ North™ NorthFlex Light Task and Light Task Plus II™ Gloves
PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life
Manufacturer: Honeywell™ NF1510XL*
Description
North NorthFlex Light Task Polyurethane Coated Nylon Gloves
- Not cut-resistant
- Polyurethane palm coating provides excellent grip and longer life
- Uncoated back design allows gloves to breathe for more comfort
- 15 gauge nylon liner has minimal shrinkage when laundered
- Cuff over-stitching is color-coded by size to aid sorting after laundering
Specifications
Palm | |
Sandy | |
automotive assembly, electronics, general maintenance work, light assembly, manufacturing, small part handling | |
Nylon | |
White |
Launderable | |
Knit | |
Polyurethane | |
X-Large | |
Navy blue |