missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ North™ NorthFlex Light Task Plus II™ Gloves
PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life
Manufacturer: Honeywell™ NFD158M
Description
North NorthFlex Light Task II Polyurethane Coated Dyneema Gloves- PU (polyurethane) palm coating is light for comfort and maximum tactility, but provides high abrasion and puncture resistance
- Glove conforms to the shape of the hand and provides minimal resistance to hand movement, thus greatly reducing fatigue
- The back of the glove is un-coated, for ventilation and wearer comfort
- Contains no reaction causing, natural rubber proteins
- Cuff over-stitching is color-coded by size to aid sorting after laundering
Specifications
Palm, wrist | |
sheet Metal handling, Metal fabrication work | |
Dyneema™ Fiber | |
White | |
2.54 cm |
Knit | |
Nitrile,Polyurethane | |
Medium | |
1 in. |