North NorthFlex Cold Grip™ Gloves
Used for work in cold conditions, such as construction, transport, forestry, snow clearing and cold storage
$7.39 - $76.45
Specifications
Color Black,Blue
Exterior Finish Textured
Glove Cuff Style Knit
Temperature (English) Min -15°F
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Material Size Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Material Size Price Quantity  
19-045-561 Honeywell Safety Products
NF11HD/8M
construction, transport, cold storage, snow clearing, forestry Vinyl Medium Pair for $7.39
Pack of 12 Pair for $81.00
17-805-010 Honeywell Safety Products
NF11HD9L
cold conditions, such as construction, transport, forestry, snow clearing and cold storage PVC (Polyvinyl Chloride) Large Dozen of 12 Pair for $76.45
Case of 6 Dozen for $458.70
17-805-011 Honeywell Safety Products
NF11HD10XL
cold conditions, such as construction, transport, forestry, snow clearing and cold storage Vinyl X-Large Dozen of 12 Pair for $76.45
Case of 6 Dozen for $458.70
Specifications
Description
  • Seamless brushed synthetic liner provides warmth and comfort in cold conditions
  • PVC coating extends to the knuckles to provide liquid protection
  • Foamed coating for excellent dry and wet grip
  • Retain flexibility at low temperatures for worker dexterity
  • Recommended for temperatures down to -15°F (-26°C)
  • Launderable for added value and longer life
Description & Specifications
Specifications
Black,Blue
Textured
Knit
-15°F