Honeywell™ North™ Low-Profile Cartridges and Cartridge/P100 Filter Combinations
Satisfy numerous industrial applications
Supplier: Honeywell™ N75003L
Description
- All North air-purifying cartridges are approved with all North air-purifying half masks and full-facepiece respirators
- Air-purifying respirators with cartridges provide respiratory protection against gases and vapors by using sorbents to purify the inhaled air
- Cartridges without filters do not remove dust, fibers, and other particles
Certifications
All North cartridges, filters, and cartridge/filter combinations are NIOSH certified to 42CFR84 standards when used with North respirator facepieces.
Compliance
NIOSH approved.
Specifications
Organic Vapor, Acid Gas Cartridge | |
Yellow | |
Industrial applications | |
18/Cs. |
NIOSH, 42CFR84 | |
Cartridge | |
Respirator |