Honeywell™ North™ Lightning™ T6550 Series Safety Eyewear
Engineered for safety officers but designed for users
Manufacturer:  Honeywell™ T65505BS
 View more versions of this product
Catalog No. 29-635-2817
Description
Description
  • High base panoramic lens for a wide unobstructed field of view
  • Bowed temples designed to fit virtually anyone
  • New Soft-Touch comfort pads
  • Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
  • The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
  • Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
Specifications
Specifications
Black
Bowed w/soft-touch pads
Anti-fog,Anti-static,Anti-UV,Scratch-resistant
Polycarbonate
Smoke
Documents