Honeywell™ North™ Lightning™ T6550 Series Safety Eyewear
Engineered for safety officers but designed for users
Manufacturer: Honeywell™ T65505B
Description
- High base panoramic lens for a wide unobstructed field of view
- Bowed temples designed to fit virtually anyone
- New Soft-Touch comfort pads
- Provided with two Flexi-Fit soft nose-pieces (regular and narrow) for comfort and adaptability
- The lenses are protected with North's exclusive 4A Coating: Anti-Fog, Anti-Scratch, Anti-UV 99.9%, Anti-Static
- Meets and/or exceeds ANSI Z87.1 and CSA Z94.3 requirements
Specifications
Black | |
Bowed w/soft-touch pads | |
Clear | |
Anti-fog,Anti-static,Anti-UV,Scratch-resistant |