Please login to your online account to display your discounted pricing

North Grip N™ Hot Mill Gloves

Heavy-duty cotton gloves ideal for workers in millwork or heavy labor

$89.24 - $89.24

Specifications

Certifications 21 CFR, USDA, FDA
For Use With (Application) automotive assembly, food processing, workers in millwork or heavy labor
Glove Cuff Style Knit
Hand Ambidextrous
Temperature (English) Max. 400°F
View More Specs
Products
Catalog Number Mfr. No. Material Size Price Quantity    

19-025-803

 
honeywell safety products
51/7147
Cotton Dozen of 12 Each for $89.24
Description & Specifications

Specifications

Certifications 21 CFR, USDA, FDA
For Use With (Application) automotive assembly, food processing, workers in millwork or heavy labor
Glove Cuff Style Knit
Hand Ambidextrous
Temperature (English) Max. 400°F
Temperature (Metric) Max 204°C

  • Provides protection against intermittent heat up to 400° F
  • Nitrile palm coating provides excellent grip and longer life
  • Knit design allows hands to breathe, unlike cut/sewn hot mill gloves
  • Available with extended cuffs for additional protection above the wrist
Steamproof version has a three layer design:
  • Inner layer of cotton provides hand comfort and wicks perspiration away from skin
  • Outer layer has excellent grip
  • Protective layer in between gloves provides resistance to steam
  • All three work together to provide a lightweight solution for working in hot, steamy applications

Ambidextrous