Please login to your online account to display your discounted pricing

North Grip N™ Gloves with Kevlar™


Nitrile-coated Kevlar gloves

Manufacturer: honeywell safety products  526647M

 View more versions of this product

Catalog No. 11-393-71A

  • / Dozen of 12 Pair
  • / Case of 12 Dozen
 

Description & Specifications

Specifications

For Use With (Application) fabricating, manufacturing, sharp part handling, sheet Metal handling
Glove Cuff Style Knit
Weight Light weight
Cut Resistance Rating Level 2
Length (English) 10 in.
Size Universal, Men's
Color Yellow, rust
Coating Material Seamless
Length (Metric) 25.4cm
Material Kevlar™
Hand Ambidextrous