missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ North™ Glove Box Gloves

Hand specific and ambidextrous styles available for comfortable fit

Manufacturer:  Honeywell™ 8Y1532A9Q

 View more versions of this product

Catalog No. 19-130-2256



Description

Description

Hand specific and ambidextrous styles available for comfortable fit

  • Butyl provides highest permeation resistance to vapors and gases, as well as exceptional
  • Resistance to a broad range of toxic chemicals, making it only suitable glove choice for many critical applications
  • Highly resistant to ketones (MEK, MIBK, acetone) and esters (tricresyl phosphate, amyl acetate, ethyl acetate)
  • Butyl has soft and flexible feel, even at low temperatures, which reduces hand fatigue and enhances worker comfort
  • Solvent based, multi-dip manufacturing process provides multiple layers of polymer (like glove within glove) which results in glove of superior quality

Black Neoprene Box Gloves

  • Manufactured from premium quality milled neoprene which provides high permeation resistance to vapors & gases
  • Resistant to a broad range of chemicals, including oils, greases, fuels and acetone
Black Butyl Box Gloves
  • Butyl provides the highest permeation resistance to vapors and gases, as well as exceptional resistance to a broad range of toxic chemicals, making it the only suitable glove choice for many critical applications
  • Highly resistant to ketones (MEK, MIBK, acetone) and esters (tricresyl phosphate, amyl acetate, ethyl acetate)
White CSM Glove Box Gloves
  • CSM provides superior resistance to ozone and oxidizing chemicals such as alcohols, alkalis and acids
  • Highly resistant to the effects of UV radiation in both artificial light and sunlight

Order Info

For chemical resistance data on these gloves, call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Also available in ambidextrous style in sizes of 9.75 in. (25cm) and 10.5 in. (27cm). North Viton™ dry box gloves are available on special order. Other sizes available include 8.5 in. (22cm), 10.5 in. (27cm), and 11.5 in. (29cm). Please call your Fisher Safety Customer Service Representative for pricing information. All gloves in pack of one pair.

Compliance

Meets EN 388-2003, EN 374-3 and EN 420-2003 standards.

Specifications

Specifications

9.75 in.
8 in.
Smooth
CSM
15mil
32 in.
25 cm
20.32 cm
Ambidextrous
8 in.
White
81.28 cm
Documents