Honeywell™ North™ Fuse™ Safety Glasses
Deliver exceptional coverage with both style and comfort
Manufacturer:  Honeywell™ UV11150801
 View more versions of this product
Catalog No. 191300840
Description
Description
Deliver exceptional coverage with both style and comfort
•    Lightweight, durable, gunmetal frame
•    Dual 9–base wrap-around lens design for full unobstructed field of vision
•    Adjustable saddle nose pads, soft temple tips
•    Durable polycarbonate lenses with scratch-resistant or Fog-Ban™ antifog coating
•    Adjustable breakaway Flexicord™ included
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 safety standards
Specifications
Specifications
Meet ANSI Z87+ and CSA Z94.3 safety standards
Break-Away Flexicord
Scratch-resistant
Gun Metal
TSR Gray