Please login to your online account to display your discounted pricing
North Emergency Escape Breathing Apparatus (EEBA)
Helps workers escape safely in an emergency
$688.50 - $688.50
Products
Catalog Number Mfr. No. Price Quantity    
17-623-9
honeywell safety products
850*
Each for $688.50
Description & Specifications
  • Low-maintenance design ensures EEBAs are ready when needed
  • Durable polyurethane hood with elastic neckband simply slips over wearer's head
  • High-visibility carrying bags are easy to locate, even in smoky conditions; illustrated instructions are clearly printed on each case
  • 360° transparent air hood has a deflector to blow incoming air onto the lens to minimize fogging
  • ON/OFF valve is easy to locate and operate
  • Pressure gauge is easy to read
Optional molded plastic storage cases are also available for easy carrying and storage. Call your Fisher Safety Customer Service Representative at 1-800-772-6733 for description of cases.
Lightweight, refillable aluminum air cylinders are approved by the Department of Transportation with a five-year hydrostatic test cycle.
NIOSH/MSHA approved for respiratory protection during escape from oxygen-deficient atmospheres, gases, and vapors at temperatures above -17°C (0°F).
Pressure-reducing valve is specially designed to reduce maintenance requirements and carries a two-year warranty.