Honeywell™ North™ Compact Air Powered Air-Purifying Respirators: HEPA Filter
For Air powered air purifying respirator systems
Manufacturer:  Honeywell™ 40HE
Catalog No. 19-134-258
Description
Description
Certifications
NIOSH approved.
Specifications
Specifications
NIOSH
HEPA Filter
Locations where movement is critical or the area to cover exceeds 300 ft., best for occasional use that may not warrant the investment in air supply system
Hepa Filter
Magenta
Blower and battery assembly, belt and backpad, flow meter, battery charger and carrying case, multiple alarm
Compact air powered air purifying respirator