Honeywell™ North™ CFR-1™ Reusable Particulate Respirators
Replaceable filters save money and reduce waste
Manufacturer:  Honeywell™ 4200S
 View more versions of this product
Catalog No. 29-635-1862
Includes: Single N95 filter for minimum 95% efficient filtration of solids and particulate aerosols free of oil.
Description
Description
Respirators combine a reusable facepiece retainer with a replaceable cup-style filter medium.
  • Exhalation valve keeps filter dry and cool and permit easier breathing
  • Provides increased wearer comfort and extends filter life; a single filter can provide all-day protection
  • Inhalation valve also allows user to perform a positive pressure seal check
  • Facepiece features a soft thermoplastic elastomer seal and an adjustable strap head harness for a secure and comfortable fit
  • “Parking mechanism” permits wearer to hang respirator from neck when not in use
  • Welding respirator shares the features of the dust mask, plus it comes with a stainless-steel mesh cover to protect wearer from heat, sparks, and slag
Order Info
Choose from basic dust mask or welding respirator.
Specifications
Specifications
NIOSH
Dust Mask
White
N95 Filter
Single N95 filter for minimum 95% efficient filtration of solids and particulate aerosols free of oil.
Thermoplastic Elastomer
Cradle
1
Blue
No
Grinding, sanding, sweeping, dusty environment
Yes
Small
Documents