Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B32410XL

 View more versions of this product

Catalog No. 11-395-44C

  • / Pair
 

Description & Specifications

Specifications

Size 10
Glove Cuff Style Beaded/Rolled
Thickness (English) 32mil
Color Black
Length (English) 14 in.
Length (Metric) 35.5cm
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
For Use With (Application) domestic preparedness, hazmat applications
Exterior Finish Smooth
Material Butyl
Hand Hand-specific