Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B3249L

 View more versions of this product

Catalog No. 11-395-44B

  • / Pair
 

Description & Specifications

Specifications

Size 9
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
Exterior Finish Smooth
Glove Cuff Style Beaded, Rolled
Hand Hand-specific
For Use With (Application) domestic preparedness, hazmat applications
Length (English) 14 in.
Length (Metric) 35.5cm
Material Butyl
Thickness 32mil
Color Black