Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B3248M

 View more versions of this product

Catalog No. 11-395-44A

  • / Pair
 

Description & Specifications

Specifications

Glove Cuff Style Beaded/Rolled
Size 8
Length (Metric) 35.5cm
Thickness (English) 32mil
Length (English) 14 in.
Color Black
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
For Use With (Application) domestic preparedness, hazmat applications
Exterior Finish Smooth
Hand Hand-specific
Material Butyl