Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B13111XXL

 View more versions of this product

Catalog No. 11-394-9E

  • / Pair
 

Description & Specifications

Specifications

Size 11
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
Exterior Finish Smooth
Glove Cuff Style Beaded, Rolled
Hand Hand-specific
For Use With (Application) Ketones, Esters, high permeation resistance to Gas or Water vapors
Length (English) 11 in.
Length (Metric) 27.9cm
Material Butyl
Thickness 13mil
Color Black