Honeywell North™ Butyl Gloves
High permeation resistance to gas or water vapors
Manufacturer:  Honeywell B1318M
 View more versions of this product
Catalog No. 11-394-9B
Specifications Description & Specifications
Specifications
Beaded/Rolled
11 in.
13mil
ANSI/ISEA, ASTM, EN, 29 CFR
Smooth
Butyl
8
Black
27.9cm
Ketones, Esters, high permeation resistance to Gas or Water vapors
Hand-specific