Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B1317S

 View more versions of this product

Catalog No. 11-394-9A

  • / Pair
 

Description & Specifications

Specifications

Size 7
Length (Metric) 27.9cm
Length (English) 11 in.
Glove Cuff Style Beaded/Rolled
Thickness (English) 13mil
Color Black
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
For Use With (Application) Ketones, Esters, high permeation resistance to Gas or Water vapors
Exterior Finish Smooth
Hand Hand-specific
Material Butyl