Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B174R10XL

 View more versions of this product

Catalog No. 11-394-8C

  • / Pair
 

Description & Specifications

Specifications

Size 10
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
For Use With (Application) Ketones, Esters, high permeation resistance to Gas or Water vapors
Glove Cuff Style Beaded, Rolled
Exterior Finish Rough grip saf
Length (Metric) 35.5cm
Material Butyl
Hand Hand-specific
Thickness 17mil
Color Black
Length (English) 14 in.