Please login to your online account to display your discounted pricing

Honeywell North™ Butyl Gloves


High permeation resistance to gas or water vapors

Manufacturer: honeywell  B174R8M

 View more versions of this product

Catalog No. 11-394-8A

  • / Pair
 

Description & Specifications

Specifications

Color Black
Glove Cuff Style Beaded/Rolled
Size 8
Length (Metric) 35.5cm
Thickness (English) 17mil
Length (English) 14 in.
Certifications ANSI/ISEA, ASTM, EN, 29 CFR
For Use With (Application) Ketones, Esters, high permeation resistance to Gas or Water vapors
Exterior Finish Rough grip saf
Material Butyl
Hand Hand-specific