Please login to your online account to display your discounted pricing

North Bulk First Aid Kits


Designed for large facilities.

Manufacturer: fisher scientific  019705-4505/EMD

 View more versions of this product

Catalog No. S98414C

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ANSI Z308.1-2003
For Use With (Application) Manufacturing settings where a wide variety of first aid items are required
Includes Metal
Sufficient For 50 Persons
Product Type First Aid Kit