Please login to your online account to display your discounted pricing

North Blue 1 Half Mask Respirators

$36.85 - $37.17

Specifications

Cartridge Type S Series Cartridge
Color Blue
Product Type Half Mask Respirator
For Use With (Application) Fire protection, government, manufacturing, medical, military, mining, pharmaceutical, steel and metals
Includes Silicone skirt, speaking diaphragm, dual sealing flange with one inner and one outer flange, rear split strap, retainer clips
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-068523

 
north safety products
B210010
Small Each for $37.17

19-068525

 
north safety products
B230010
Large Each for $36.85
Description & Specifications

Specifications

Cartridge Type S Series Cartridge
Color Blue
Product Type Half Mask Respirator
For Use With (Application) Fire protection, government, manufacturing, medical, military, mining, pharmaceutical, steel and metals
Includes Silicone skirt, speaking diaphragm, dual sealing flange with one inner and one outer flange, rear split strap, retainer clips
Material Silicone