Honeywell North A900 Series Safety Eyewear
Ultra-lightweight eyewear features superior style
$3.57 - $3.57
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003 standards.
Frame Color Black
Lens Material Polycarbonate
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Lens Tint Lens Type Price Quantity  
Catalog Number Mfr. No. Lens Tint Lens Type Price Quantity  
19-160-1201 Honeywell
A901CSA
Clear Anti-fog Each for $3.57
Specifications
Description
  • Sporty, eight-base wraparound dual lens provides style, comfort and protection
  • Temples with padded inserts offer a comfortable fit
  • Available with scratch-resistant hardcoat or Fog-Ban™ antifog coating
  • Non-slip rubber nosepiece
  • Black frames
Compliance
Meets ANSI™ Z87.1-2010 standards.
Description & Specifications
Specifications
Meets ANSI Z87.1-2003 standards.
Black
Polycarbonate