North™ A700 Series Safety Glasses
Sporty styling provides fashionable fit for the everyday wearer
$1.98 - $31.98
Specifications
Certifications/Compliance Meets or exceed ANSI Z87.1-2003 standards.
Lens Material Polycarbonate
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Frame Color Lens Tint Lens Type Price Quantity  
Catalog Number Mfr. No. Frame Color Lens Tint Lens Type Price Quantity  
19-160-0217 Accessories North Safety Products
UVA700
Clear Clear Scratch-resistant Each for $1.98
Pack of 10 Each for $19.20
19-160-0218 North Safety Products
UVA701
Gray Gray Scratch-resistant Each for $2.19
Pack of 10 Each for $21.30
19-160-0220 Accessories North Safety Products
UVA703
Gray Blue Mirror Scratch-resistant Pack of 10 Each for $31.98
19-160-0221 North Safety Products
A704
Gray Silver Mirror Scratch-resistant Pack of 10 Each for $31.98
19-160-0222 Accessories North Safety Products
UVA705
Clear Clear Scratch-resistant Each for $2.59
Pack of 10 Each for $25.10
19-160-0219 North Safety Products
A702
Clear Amber Scratch-resistant Each for $2.19
Pack of 10 Each for $21.30
Specifications
Description
  • Nine-base polycarbonate hardcoated lenses provide 180° field of distortion-free vision; block 99.9% UV
  • Lightweight design with sport temples and soft temple-tip pads for secure, wraparound fit
  • Molded universal nose bridge for comfortable fit
Compliance
Meet or exceed ANSI™ Z87.1-2010 standards.
Description & Specifications
Specifications
Meets or exceed ANSI Z87.1-2003 standards.
Polycarbonate