Honeywell™ North™ A400 Series Safety Glasses
Sporty, lightweight safety glasses with one-piece wraparound lens
Supplier: Honeywell™ UVA401
Description
- Single lens and frame made of impact-resistant polycarbonate
- Comfortable, wraparound design provides excellent peripheral vision protection without the need for sideshields
- Feature antifog coating
Compliance
Meet ANSI™ Z87.1-2003 and CSA Z94.3 standards.
Specifications
Meet ANSI Z87.1-2003 and CSA Z94.3 standards. | |
Gray | |
Gray |