Honeywell™ North™ 4000 Series Full Facepiece Respirators
The 4000 Full Facepiece respirator is an industry classic in black and with welding attachment options.
$409.21 - $409.21
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-003-005
View Documents
Honeywell™
420079
Small (Standard)
Each for $409.21
Description
Description
  • Durable silicone facepiece has wide sealing flanges for comfort and seal integrity with a minimum of headstrap pressure.
  • Single-curve polycarbonate lens provides wearer with an unobstructed wide-angle view.
  • Inside surface features an antifog coating effective in temperatures as low as -31.6°C (-25°F), providing unimpaired vision in even the coldest climates.
  • Offset browguard and special surface coating protect lens from scratches.
  • Four-point cradle headstrap maintains a secure fit with comfortable tension.
  • An advanced speaking diaphragm allows clear communication with less annoying resonance.
With minor conversions, the full facepiece can be used with other Survivair respirators. Meets NIOSH 42CFR84.
SDS