Please login to your online account to display your discounted pricing

NorLake™ Scientific Select Series Temperature Stability Test Chambers w/Glass Doors

Advanced engineered design for optimal storage temperature environments

Specifications

Certifications/Compliance UL, C-UL
Display LED
Refrigerant R-134a
View More Specs
Products
Catalog Number Mfr. No. Capacity Electrical Requirements No. of Doors Price Quantity    

22-650-828

 
nor lake inc
NSRI241WSG0
684L (24.1 cu. ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 1 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-839

 
nor lake inc
NSRI803WSG5
2269L (79.9 cu. ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 3 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-838

  Accessories
nor lake inc
NSRI803WSG4
2269L (79.9 cu. ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 3 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-837

 
nor lake inc
NSRI803WSG0
2269L (79.9 cu. ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 3 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-835

  Accessories
nor lake inc
NSRI522WSG4
1473L (51.9 cu. ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 2 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-836

 
nor lake inc
NSRI522WSG5
1473L (51.9 cu. ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 2 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-834

 
nor lake inc
NSRI522WSG0
1473L (51.9 cu. ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 2 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-832

  Accessories
nor lake inc
NSRI331WSG4
940L (33.1 cu.ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 1 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-833

 
nor lake inc
NSRI331WSG5
940L (33.1 cu.ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 1 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-831

 
nor lake inc
NSRI331WSG0
940L (33.1 cu.ft.) 115V, 1PH, 60Hz, 14.7A; Nema plug: 5-20P; Nema Receptacle: 5-20R 1 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-830

 
nor lake inc
NSRI241WSG5
684L (24.1 cu. ft.) 230V, 1PH, 50Hz, 7.5A; Power Inlet (IEC 60320) module 1 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

22-650-829

  Accessories
nor lake inc
NSRI241WSG4
684L (24.1 cu. ft.) 208-230V, 1PH, 60Hz, 7.5A; Nema plug: 6-15P; Nema Receptacle: 6-15R 1 CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Certifications/Compliance UL, C-UL
Display LED
Refrigerant R-134a