Please login to your online account to display your discounted pricing

Norketamine hydrochloride, Tocris

Potent, non-competitive NMDA antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Norketamine hydrochloride
Assay Percent Range >99%
Molecular Formula C12H14ClNO·HCl
Formula Weight 260.16
Synonym 2-Amino-2-(2-chlorophenyl)cyclohexanone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

197010

 
r & d systems
RDS197010
10mg Each for $181.19

197050

 
r & d systems
RDS197050
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 260.16
Quantity 10mg
Chemical Name or Material Norketamine hydrochloride
Assay Percent Range >99%
Molecular Formula C12H14ClNO·HCl
CAS 79499-59-5
Synonym 2-Amino-2-(2-chlorophenyl)cyclohexanone hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature