missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Norcamphor, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129250250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Norcamphor | |
98.5 | |
88°C to 91°C | |
33g/mL | |
33°C | |
99% | |
C7H10O | |
25 g | |
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one | |
KPMKEVXVVHNIEY-UHFFFAOYSA-N | |
bicyclo[2.2.1]heptan-3-one | |
10345 | |
99% |
497-38-1 | |
100.0 | |
Colorless to White | |
168°C to 172°C | |
Authentic | |
Plastic bottle | |
MFCD00074823 | |
07, 57 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
C1CC2CC1CC2=O | |
110.16 | |
110.16 | |
Adhering Crystals |
Chemical Identifiers
497-38-1 | |
110.16 | |
KPMKEVXVVHNIEY-UHFFFAOYSA-N | |
10345 | |
C1CC2CC1CC2=O |
C7H10O | |
MFCD00074823 | |
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-3-one | |
bicyclo[2.2.1]heptan-3-one |
Safety and Handling
GHS H Statement
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 207-846-1
RUO – Research Use Only