missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Norcamphor, 98+% Alfa Aesar™

Used as a building block in organic synthesis and also acts as a precursor to norborneols.

$43.95 - $128.31

Chemical Identifiers

CAS 497-38-1
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00074823
InChI Key KPMKEVXVVHNIEY-UHFFFAOYSA-N
Synonym norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-2-one, bicyclo 2.2.1 heptan-3-one
PubChem CID 10345
IUPAC Name bicyclo[2.2.1]heptan-3-one
SMILES C1CC2CC1CC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0149106 View Documents Alfa Aesar
L0149106
5g Each for $43.95
 
AAL0149114 View Documents Alfa Aesar
L0149114
25g Each for $128.31
 
Description

Description

Applications
Norcamphor is used as a building block in organic synthesis. It is also used as a precursor to norborneols.

Solubility
Insoluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Heat and light sensitive.
Specifications

Chemical Identifiers

497-38-1
110.156
KPMKEVXVVHNIEY-UHFFFAOYSA-N
10345
C1CC2CC1CC2=O
C7H10O
MFCD00074823
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-2-one, bicyclo 2.2.1 heptan-3-one
bicyclo[2.2.1]heptan-3-one

Specifications

33°C (91°F)
92°C to 97°C
497-38-1
MFCD00074823
1209657
Insoluble in water. Soluble in methanol.
C1CC2CC1CC2=O
110.156
110.16
Norcamphor
172°C to 174°C
5g
C7H10O
UN1325
norcamphor, 2-norbornanone, bicyclo 2.2.1 heptan-2-one, norcampher, 2-oxonorbornane, norbornan-2-one, 2,5-methanocyclohexanone, 8,9,10-trinorbornan-2-one, bicyclo 2.2.1 heptan-2-one, bicyclo 2.2.1 heptan-3-one
KPMKEVXVVHNIEY-UHFFFAOYSA-N
bicyclo[2.2.1]heptan-3-one
10345
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : RB7680000

TSCA : TSCA