Please login to your online account to display your discounted pricing

Alfa Aesar™ (Norbornadiene)rhodium(I) 2,4-pentanedionate, Rh 34.9%

Chemical Identifiers

Chemical Name or Material (Norbornadiene)rhodium(I) 2,4-pentanedionate
CAS 32354-50-0
Formula Weight 294.15
MDL Number MFCD00075071
Synonym Acetylacetonato(norbornadiene)rhodium(I); Ir(acac)(nbd)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4672703

 
alfa aesar™
4672703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 294.15
Melting Point 176° to 178°C
Chemical Name or Material (Norbornadiene)rhodium(I) 2,4-pentanedionate
Concentration Rh 34.9%
Quantity 1g
CAS 32354-50-0
MDL Number MFCD00075071
Color Yellow
Synonym Acetylacetonato(norbornadiene)rhodium(I); Ir(acac)(nbd)
Physical Form Crystalline Solid