missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nor-Lake™ Scientific Premier Glass Door Freezer

NorLake™ Premier Glass Door Freezer provides optimal storage temperature environments necessary for a wide range of life science, pharmacy, biological, medical, clinical and industrial applications.

Manufacturer:  Nor-Lake™ Scientific NSXF211WWG/0*

 View more versions of this product

Catalog No. 22-651-127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: Power cord and plug

Description

Description

  • Digital LED microprocessor temperature controller with Hi/Lo audible and visual alarm
  • Audible and visual Hi/Lo alarm, remote alarm contacts
  • Continuous product temperature display, product sensor with glycerin bottle, power supply switch
  • Exterior cabinet front, sides and back are painted white
  • Exterior cabinet top and bottom are galvanized steel, interior cabinet liner painted white
  • Cabinet is foamed-in-place with CFC-free high-density polyurethane foam insulation
  • Glass door(s), full size, double pane, with anti-condensate heater wires
  • Heavy-duty door pivot hinges, centered key door locks (two keys)
  • Pull door handle(s)
  • Four casters (two locking)
  • Three epoxy coated wire shelves per door, perimeter anti-condensate door opening heater wires, magnetic door gasket
  • Switch-activated interior light

Order Info

Options include legs in lieu of casters, stainless-steel drawers (eight per door), wire basket drawers (six per door), extra shelf, stainless-steel exterior, stainless-steel interior, 4-20ma output, temperature chart recorder with chart paper, sensor access port (1 in. [2.5cm] dia.), export crating.

Certifications

UL listed

Warranty

18 month parts and labor, 5 year compressor (US and Canada) 18 months parts (International)

Specifications

Specifications

CFC-free foamed-in HDPF
Upright
cUL,UL
594.6 L
115 V
14.9 A
35.5 x 27.16 x 81.5 in. (90 x 69 x 207 cm)
485 lb.
LED
White
Microprocessor
76 cm
59 in.
149 cm
90 cm
59 cm
27.16 in.
3
-10°C to -25°C
21 cu. ft.
115 V 60 Hz
60 Hz
NEMA 5-15
30 x 23.5 x 59 in. (76 x 59 x 149 cm)
219.9 kg
R-404A
Power cord and plug
30 in.
81.5 in.
207 cm
35.5 in.
23.5 in.
Automatic
69 cm
SDS
Documents
Accessories