Nor-Lake™ Scientific Blood Bank Refrigerators
Provides energy efficient, convenient, safe and reliable performance for optimal storage
Specifications
Certifications/Compliance UL, cUL
Color White
Cabinet Material Galvanized Steel
Defrost Automatic
Display LCD
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Electrical Requirements Capacity Temperature Range (Metric) Price Quantity  
Catalog Number Mfr. No. Electrical Requirements Capacity Temperature Range (Metric) Price Quantity  
36-100-5518 Nor Lake Inc
NSBR241WWG0
115V 60Hz 24 cu. ft., 680L 1° to 6°C N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-650-655 Accessories Nor Lake Inc
NSBR241WWG/5
230V 50Hz 24 cu. ft., 680L 1° to 6°C N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
22-651-102 Nor Lake Inc
NSBR331WWG/0
115V 60Hz 33 cu. ft., 934L 1° to 6°C N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
  • Capacities from 24 to 80 cu. ft.
  • Operating temperature 1° to 6°C (factory preset at 4°C)
  • Microprocessor controller with 4x20 character LCD digital display control panel with message center
  • 100-event alarm logging, including date/time stamp and product temperatures; stored in nonvolatile flash memory
  • Two levels of user password protection (setpoints and parameters)
  • Audible and visual high/low product temperature; manually adjustable alarm volume
  • Door-ajar alarm with adjustable delay time period
  • Remote alarm contacts
  • Air-temperature probe with high/low alarms
  • Power failure alarm
  • Integrated high/low alarm tests
  • Two content temperature probes with high/low alarms
  • Continuous product temperature display
  • Two product sensors with glycerin bottles
  • Power supply switch
  • Temperature chart recorder and chart paper
  • 2 to 10VDC output for product temperature (selectable to air temperature)
  • Air and product digital calibration
  • Real-time clock with date and time display
  • Door opening counter with 24-hour automatic reset
  • Low battery test switch with buzzer
  • Available with one, two, or three glass doors
  • Interior cabinet liner and exterior cabinet front, sides and back are painted white; exterior back and bottom are galvanized steel
  • Foamed-in-place, high-density CFC-free polyurethane insulation
  • Automatic defrost and condensate evaporation
  • Condenser/compresser run time, fan motor run time, and defrost heater time monitors
  • Heavy-duty doors feature pull-door handles, heavy-duty pivot hinges, center key door locks, and magnetic door gaskets
  • Perimeter anti-condensate door grame (temperature selectable) heater wires
  • Switch-activated interior lighting
  • Air-cooled condensing unit
  • Automatic condensate evaporator
  • Eight solid stainless-steel drawers per door
  • Switch-activated fluorescent interior lighting
  • Top-mounted forced air refrigeration system
  • Four casters (two locking) on one- and two-door models; six casters (three locking on three-door models
  • 1 in. dia. sensor access port
Available options include extra shelves, wire basket drawer, stainless steel interior/exterior, RS-485 communication port, and 4-20ma output. Contact your Fisher Scientific™ representative for more information.
Compliance
UL/CUL listed; CE marked; CEC (California Energy Commission) rated
Warranty
18 months parts and labor, 5 years compressor (US and Canada); 18 months parts (International)
Description & Specifications
Specifications
UL, cUL
Galvanized Steel
LCD
CFC-free foamed-in Polyurethane
R-134A
18 Months
White
Automatic
1
RS485
Microprocessor