Please login to your online account to display your discounted pricing

nor-Binaltorphimine dihydrochloride, Tocris

Standard κ selective antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material nor-Binaltorphimine dihydrochloride
Assay Percent Range >98%
Molecular Formula C40H43N3O6·2HCl
Formula Weight 734.72
Synonym 17,17'-(Dicyclopropylmethyl)-6,6',7,7'-6,6'-imino- 7,7'-binorphinan-3,4',14,14'-tetrol dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

034750

 
r & d systems
RDS034750
50mg Each for $844.73

034710

 
r & d systems
RDS034710
10mg Each for $201.36
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 734.72
Chemical Name or Material nor-Binaltorphimine dihydrochloride
CAS 113158-34-2
Assay Percent Range >98%
Molecular Formula C40H43N3O6·2HCl
Synonym 17,17'-(Dicyclopropylmethyl)-6,6',7,7'-6,6'-imino- 7,7'-binorphinan-3,4',14,14'-tetrol dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature