Please login to your online account to display your discounted pricing

nor-Binaltorphimine dihydrochloride, Tocris


Standard κ selective antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS034750

 View more versions of this product

Catalog No. 034750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 734.72
Chemical Name or Material nor-Binaltorphimine dihydrochloride
CAS 113158-34-2
Assay Percent Range >98%
Molecular Formula C40H43N3O6·2HCl
Synonym 17,17'-(Dicyclopropylmethyl)-6,6',7,7'-6,6'-imino- 7,7'-binorphinan-3,4',14,14'-tetrol dihydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material nor-Binaltorphimine dihydrochloride
Assay Percent Range >98%
Molecular Formula C40H43N3O6·2HCl
Formula Weight 734.72
Synonym 17,17'-(Dicyclopropylmethyl)-6,6',7,7'-6,6'-imino- 7,7'-binorphinan-3,4',14,14'-tetrol dihydrochloride
Accessories

Store at Room Temperature