missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonylcyclohexane, TCI America™

$168.06 - $482.78

Chemical Identifiers

CAS 2883-02-5
Molecular Formula C15H30
Molecular Weight (g/mol) 210.41
MDL Number MFCD00058955
InChI Key CLMFECCMAVQYQA-UHFFFAOYSA-N
Synonym 1-Cyclohexylnonane
PubChem CID 17900
IUPAC Name nonylcyclohexane
SMILES CCCCCCCCCC1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N03795ML View Documents TCI America
N03795ML
5mL Each for $168.06
 
N037925ML View Documents TCI America
N037925ML
25mL Each for $482.78
 
Specifications

Chemical Identifiers

2883-02-5
210.41
CLMFECCMAVQYQA-UHFFFAOYSA-N
17900
CCCCCCCCCC1CCCCC1
C15H30
MFCD00058955
1-Cyclohexylnonane
nonylcyclohexane

Specifications

282°C
5mL
C15H30
1-Cyclohexylnonane
CCCCCCCCCC1CCCCC1
210.41
210.41
≥98.0% (GC)
Colorless
2883-02-5
MFCD00058955
CLMFECCMAVQYQA-UHFFFAOYSA-N
nonylcyclohexane
17900
Clear Liquid at 20°C
Nonylcyclohexane
Safety and Handling

Safety and Handling

TSCA : Yes