missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nonyl beta-D-glucopyranoside, 98%, Thermo Scientific Chemicals
$467.11 - $1539.05
Chemical Identifiers
CAS | 69984-73-2 |
---|---|
Molecular Formula | C15H30O6 |
Molecular Weight (g/mol) | 306.399 |
MDL Number | MFCD00063300 |
InChI Key | QFAPUKLCALRPLH-UXXRCYHCSA-N |
Synonym | nonyl beta-d-glucopyranoside, b-nonylglucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy tetrahydro-2h-pyran-3,4,5-triol, b-d-glucopyranoside, nonyl, nonyl-beta-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy oxane-3,4,5-triol, n-nonyl-beta-d-glucopyranoside, bng, beta-d-glucopyranoside, nonyl, 4gby |
PubChem CID | 155448 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol |
SMILES | CCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6600903
|
Thermo Scientific Chemicals
J6600903 |
1 g |
Each for $467.11
|
|
|||||
AAJ6600906
|
Thermo Scientific Chemicals
J6600906 |
5 g |
Each for $1,539.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69984-73-2 | |
306.399 | |
QFAPUKLCALRPLH-UXXRCYHCSA-N | |
155448 | |
CCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O |
C15H30O6 | |
MFCD00063300 | |
nonyl beta-d-glucopyranoside, b-nonylglucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy tetrahydro-2h-pyran-3,4,5-triol, b-d-glucopyranoside, nonyl, nonyl-beta-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy oxane-3,4,5-triol, n-nonyl-beta-d-glucopyranoside, bng, beta-d-glucopyranoside, nonyl, 4gby | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol |
Specifications
69984-73-2 | |
C15H30O6 | |
1 g | |
nonyl beta-d-glucopyranoside, b-nonylglucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy tetrahydro-2h-pyran-3,4,5-triol, b-d-glucopyranoside, nonyl, nonyl-beta-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-nonyloxy oxane-3,4,5-triol, n-nonyl-beta-d-glucopyranoside, bng, beta-d-glucopyranoside, nonyl, 4gby | |
CCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O | |
306.399 | |
306.4 | |
Powder |
White | |
MFCD00063300 | |
12168 | |
QFAPUKLCALRPLH-UXXRCYHCSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol | |
155448 | |
98% | |
Nonyl beta-D-glucopyranoside |
Safety and Handling
EINECSNumber : 615-042-0
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only