missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Drierite™ Drying Desiccants, W.A. Hammond
Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Supplier: W A Hammond Drierite Co HDC13001
Description
Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
- All purpose desiccant made of anhydrous calcium sulfate
- Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
- Dries organic liquids to low ppm levels
- Commonly used in desiccators, desiccator cabinets, in-line air and gas; dryers, glove boxes, sealed chambers
- Useful for water removal from liquid reaction mixtures and multi-component;gas streams
- Neutral, stable, constant in volume, and inert except toward water
- Insoluble in organic solvents
- Absorbs moisture at temperatures up to 66°C
Specifications
Calcium Sulfate | |
Calcium Sulfate | |
CaO4S | |
8 Mesh | |
Abosrbent | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 |
7778-18-9 | |
1 lb. | |
MFCD00010912 | |
Glass Bottle | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium sulfate | |
24497 |
Chemical Identifiers
CaO4S | |
MFCD00010912 | |
24497 | |
calcium sulfate |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
CHEBI:31346 | |
[Ca++].[O-]S([O-])(=O)=O |