missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
W.A. Hammond Drierite™ Drying Desiccants
Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Manufacturer: W.A. Hammond HDC12005
Description
Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
- All purpose desiccant made of anhydrous calcium sulfate
- Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
- Dries organic liquids to low ppm levels
- Commonly used in desiccators, desiccator cabinets, in-line air and gas; dryers, glove boxes, sealed chambers
- Useful for water removal from liquid reaction mixtures and multi-component;gas streams
- Neutral, stable, constant in volume, and inert except toward water
- Insoluble in organic solvents
- Absorbs moisture at temperatures up to 66°C
Specifications
Calcium Sulfate | |
5 lb. | |
Nonindicating; 6 Mesh; 5 lb. | |
[O-]S(=O)(=O)[O-].[Ca+2] | |
136.134 | |
CHEBI:31346 |
7778-18-9 | |
CaO4S | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium;sulfate | |
24497 |
Chemical Identifiers
7778-18-9 | |
136.134 | |
24497 | |
calcium;sulfate |
CaO4S | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
CHEBI:31346 | |
[O-]S(=O)(=O)[O-].[Ca+2] |