missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Drierite™ Drying Desiccants, W.A. Hammond

Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.

Manufacturer:  W A Hammond Drierite Co HDC12001

 View more versions of this product

Catalog No. 07-577-2A



Description

Description

Dries air and gases to lowest residual moisture level. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.

  • All purpose desiccant made of anhydrous calcium sulfate
  • Dries air and gases to the lowest residual moisture level attainable with a safe, easy to use desiccant (-100°F dew point)
  • Dries organic liquids to low ppm levels
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas; dryers, glove boxes, sealed chambers
  • Useful for water removal from liquid reaction mixtures and multi-component;gas streams
  • Neutral, stable, constant in volume, and inert except toward water
  • Insoluble in organic solvents
  • Absorbs moisture at temperatures up to 66°C
Specifications

Specifications

Calcium Sulfate
1 lb.
Nonindicating; 6 Mesh, 1 lb.
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium;sulfate
24497
7778-18-9
CaO4S
Abosrbent, Desiccant
[O-]S(=O)(=O)[O-].[Ca+2]
136.134
CHEBI:31346

Chemical Identifiers

7778-18-9
136.134
Abosrbent, Desiccant
CHEBI:31346
[O-]S(=O)(=O)[O-].[Ca+2]
CaO4S
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
24497
calcium;sulfate
SDS
Documents
Product Certifications