missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals
$181.04 - $247.79
Chemical Identifiers
CAS | 9002-93-1 |
---|---|
Molecular Formula | C16H26O2 |
Molecular Weight (g/mol) | 250.38 |
MDL Number | MFCD00132505 |
InChI Key | JYCQQPHGFMYQCF-UHFFFAOYSA-N |
Synonym | Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether |
PubChem CID | 5590 |
IUPAC Name | 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol |
SMILES | CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AAJ19628AP
|
Thermo Fisher Scientific
J19628AP |
500 mL | Plastic bottle |
Each for $181.04
|
|
||||
AAJ19628K2
|
Thermo Fisher Scientific
J19628K2 |
1 L | Plastic bottle |
Each for $247.79
|
|
||||
Chemical Identifiers
9002-93-1 | |
250.38 | |
JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
5590 | |
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 |
C16H26O2 | |
MFCD00132505 | |
Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether | |
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol |
Specifications
9002-93-1 | |
>250°C | |
1.061 | |
MFCD00132505 | |
Igepal CA to 630,Polyethylene glycol tert to octylphenyl ether | |
JYCQQPHGFMYQCF-UHFFFAOYSA-N | |
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol | |
5590 | |
Plastic bottle | |
Nonidet™ P40 Substitute |
Viscous Liquid | |
∼5°C | |
C16H26O2 | |
500 mL | |
>110°C | |
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1 | |
250.38 | |
ca 625 | |
1.061 |
Safety and Handling
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
RTECSNumber : MD0907600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only