missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonenylsuccinic Anhydride (mixture of branched chain isomers), TCI America™

$56.53 - $386.53

Chemical Identifiers

CAS 28928-97-4
Molecular Formula C13H20O3
MDL Number MFCD00014552
Synonym Isononenylsuccinic Anhydride
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0350500G View Documents TCI America
N0350500G
500g Each for $386.53
 
N035025G View Documents TCI America
N035025G
25g Each for $56.53
 
Specifications

Chemical Identifiers

28928-97-4
C13H20O3
MFCD00014552
Isononenylsuccinic Anhydride

Specifications

Yellow
28928-97-4
MFCD00014552
224.30
≥90.0% (T)
500g
C13H20O3
Isononenylsuccinic Anhydride
Clear Liquid at 20°C
Nonenylsuccinic Anhydride (mixture of branched chain isomers)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0852

TSCA : Yes