missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonenylsuccinic anhydride, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1760730

 View more versions of this product

Catalog No. AAA1760730



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nonenylsuccinic anhydride
1.032
146°C (295°F)
MFCD00014552
Moisture sensitive
96%
28928-97-4
126°C to 130°C (0.15 mmHg)
C13H20O3
250 g
224.3

Chemical Identifiers

28928-97-4
C13H20O3
MFCD00014552
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 249-317-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only