Please login to your online account to display your discounted pricing

Nonaqueous Buffer Stock Solution A, for Acid Number of Petroleum Products, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Nonaqueous Buffer Stock Solution A
Name Note For Acid Number of Petroleum Products
CAS 67-63-0
CAS Min % 93.0
CAS Max % 97.0
CAS 108-75-8
CAS Min % 2.9
CAS Max % 3.01
CAS 7647-01-0
CAS Min % 1.76
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5462-16

 
ricca chemical company
RI546216
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5462-32

  SDS
ricca chemical company
RI546232
Glass amber 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Nonaqueous Buffer Stock Solution A
Name Note For Acid Number of Petroleum Products
CAS 67-63-0
CAS Min % 93.0
CAS Max % 97.0
CAS 108-75-8
CAS Min % 2.9
CAS Max % 3.01
CAS 7647-01-0
CAS Min % 1.76
CAS Max % 1.83
Physical Form Liquid
Packaging Glass amber
Color Clear light tan
Quantity 500mL